Learn More
Description
PA1-4951 detects the HIV-1 p24 protein in HIV samples. PA1-4951 has been successfully used in Western blot and ELISA procedures. By Western blot this antibody detects a ~24 kDa protein, a ~55 kDa protein, and a ~41 kDa protein, corresponding respectively to HIV-p24 and to its precursors p55 and p41, in HIV samples. PA1-4951 immunogen corresponds to the recombinant protein which was over-expressed in E. coli with a plasmid carrying the Gag p24 coding region of the HIV-1 virus. Recombinant HIV-1 Gag p24 protein (Cat. # RP-4916 or RP-4917) can be purchased for control experiments in Western blot assays.
Specifications
Specifications
| Antigen | HIV1 p24 |
| Applications | Dot Blot, ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 0.09% sodium azide |
| Gene Alias | HIV p24; HIV1gp1 |
| Host Species | Rabbit |
| Immunogen | Purified full-size recombinant HIV1 p24, subtype B expressed in E. coli. |
| Quantity | 100 μL |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
