Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIV1 p55 Monoclonal Antibody (3A1)

Catalog No. 01670321
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-321 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-321 Supplier Invitrogen™ Supplier No. MA17587
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7587 detects HIV1 from virus samples. MA1-7587 has been successfully used in ELISA, Western blot, and immunoprecipitation procedures. The MA1-7587 immunogen is a purified recombinant HIV1 p55/17 protein.

HIV-1 Gag p55 is a precursor protein of several proteins that form the core structure of AIDS virus, indispensable to its reproduction. This protein is digested by HIV-1 protease, first into intermediate products p41 and p15. Then p41 is digested into matrix protein p17 and capsid protein p24. Protein p15 is further digested into nucleocapsid protein p7 and to p6 and p1, both of unknown function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 p55
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide; pH 7.2
Gene Alias HIV p55
Host Species Mouse
Immunogen Purified recombinant HIV1 p55/17 protein.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.