Learn More
Invitrogen™ HIV1 Protease Monoclonal Antibody (1696)

Description
This antibody recognizes free N-terminus of mature HIV protease (HIV-1 and HIV-2), an enzyme that hydrolyzes polyproteins of HIV viruses into functional proteins. The antibody 1696 does not react with the precursor. Functional Application: Inhibition.
Specifications
Specifications
| Antigen | HIV1 Protease |
| Applications | ELISA, Functional Assay, Western Blot |
| Classification | Monoclonal |
| Clone | 1696 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | HIV protease; HIV1gp1 |
| Host Species | Mouse |
| Immunogen | Bacterially expressed full-length HIV-1 protease. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.