Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIV1 Rev Group M Monoclonal Antibody (10)
GREENER_CHOICE

Catalog No. MA171514 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA171514 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA171514 Supplier Invitrogen™ Supplier No. MA171514
  • $639.00
    $506.65 / Each of 1

    Save $132.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This antibody reacts with HIV-1 Rev subtype B. It has been mapped to amino acids 34-40 (TRGARRN - subtype B a.a. consensus). This clone may also be referred to as 10 (03-002).

The HIV-1 Rev (regulator of expression of virion) protein is essential for the replication of HIV-1, especially in group M, which is the predominant and most widespread group of HIV-1 responsible for the global HIV epidemic. The Rev protein is encoded by the rev gene and plays a critical role in the nuclear export of unspliced and singly spliced viral mRNAs, which is crucial for the expression of structural proteins and the production of new virions. Structurally, Rev consists of an arginine-rich motif that allows it to bind to the Rev Response Element (RRE) within the viral RNA. This interaction facilitates the recruitment of the nuclear export receptor CRM1, which, along with other cofactors, translocates the RNA out of the nucleus. Functional disruption of Rev or its interaction with RRE significantly impedes HIV-1 replication, making these interactions potential targets for antiretroviral therapies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 Rev Group M
Applications ELISA, Western Blot, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias HIV group M; HIV-1 Rev
Host Species Mouse
Immunogen Recombinant Rev protein of HIV-1 BRV purified from E. coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.