Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIV1 Tat Monoclonal Antibody (7)
GREENER_CHOICE

Catalog No. MA171509 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA171509 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA171509 Supplier Invitrogen™ Supplier No. MA171509
  • $665.00
    $506.65 / Each of 1

    Save $158.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This antibody reacts with HIV-1 Tat BH10, IIIB strains. This antibody reacts with the rTat consensus sequences of HIV-1 subtypes A (PNLEPWH), B (PRLEPWK) and C (PNLEPWN). It has been mapped to amino acids 6-12 (PRLEPWK - B subtype a.a. consensus). MA1-71509 can be used for chromatin immunoprecipitation (ChIP) analysis of HIV1 Tat. This clone may also be referred to as 7 (02-002).

HIV-1 Tat ('Trans-Activator of Transcription') consists of 86 to 101 amino acids depending on the subtype. Tat is critical to HIV reproduction by way of compensating for a defect in its genome: the HIV RNA initially has a hairpin-structured portion which prevents full transcription from occurring. However, a small number of RNA transcripts will be made, which allow the Tat protein to be produced. Tat then binds to and phosphorylates cellular factors, eliminating the effect of the hairpin RNA structure and allowing transcription of the HIV RNA. This increases the rate of transcription, providing a positive feedback cycle. The protein is released by infected cells in culture, and is found in the blood of HIV-1 infected patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV1 Tat
Applications ChIP Assay, ELISA, Western Blot
Classification Monoclonal
Clone 7
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias HIV-1 Tat
Host Species Mouse
Immunogen Recombinant Tat protein of HIV-1 BH10 purified from E. coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.