Learn More
Abcam HIV2 Gp36 Peptide, Synthetic, >95% Purity, 500 μg
Description
- Protein Name: HIV2 gp36
- Type: Synthetic
- Purity: >95%
- Tested Applications: WB, ELISA
- Form: Liquid
- Concentration: 1 mg/mL
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Blue Ice |
| Substrate Properties | Peptide-based Substrate |
| Target Molecule | HIV2 gp36 |
| Content And Storage | Store at -20°C. |
| Label or Dye | Unconjugated |
| Concentration | 1 mg/mL |
| Form | Liquid |
| Quantity | 500 μg |
| For Use With (Application) | Western Blot, ELISA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.