Learn More
PIP™ HL Bouton Softside Goggle
Provides protection from flying particles and debris. Ideal for general manufacturing and anywhere eye protection is needed.
Supplier:  PIP™ 2485290300B
 View more versions of this product
Catalog No. 19-069150
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Solid, non-ventilated frame
  • Soft vinyl body molds snugly to the face without pressure or irritation
  • Fits comfortably over most prescription glasses
  • Extended face flanges close the gaps over the nose and cheekbone areas, providing excellent protection
  • Clear blue transparent frame to prevent yellowing
  • Clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
  • Economical style intended to be used by visitors to a facility, or other short-term applications
Specifications
Specifications
Scratch-resistant
Goggles
Non-vent
Reusable
Vinyl
Clear
Provides protection from chips and flying particles
Vinyl
Polycarbonate
UV400
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.