Learn More
PIP™ HL Bouton Softside Goggle
Catalog No. 19069151 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Clear
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Coating
19-069151 Clear Anti-fog, Scratch-resistant
19-069150 Clear Scratch-resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-069151 Supplier PIP™ Supplier No. 2485290400B
  • $1,467.00
    $882.65 / Case of 144
    Save $584.35
    40% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Provides protection from flying particles and debris. Ideal for general manufacturing and anywhere eye protection is needed.
  • Solid, non-ventilated frame
  • Soft vinyl body molds snugly to the face without pressure or irritation
  • Fits comfortably over most prescription glasses
  • Extended face flanges close the gaps over the nose and cheekbone areas, providing excellent protection
  • Clear blue transparent frame to prevent yellowing
  • Clear, crisp and distortion free visibility while meeting the requirement standards that help provide protection from projectiles and impact in general use applications
  • Economical style intended to be used by visitors to a facility, or other short-term applications
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Goggles
For Use With (Application) Provides protection from chips and flying particles
Ventilation Details Non-vent
Material Vinyl
Disposable Reusable
Lens Material Polycarbonate
Frame Material Vinyl
UV Protection UV400
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.