Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-A/HLA-B/HLA-C, Mouse anti-Human, Clone: MEM-123, Abnova™

Catalog No. 89999175 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-175 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-175 Supplier Abnova Corporation Supplier No. MAB16375
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Specifications

Antigen HLA-A/HLA-B/HLA-C
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone MEM-123
Conjugate Unconjugated
Dilution ELISA Flow Cytometry (4 ug/mL) Immunohistochemistry (Frozen sections) (10 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 8.0 (0.09% sodium azide).
Host Species Mouse
Immunogen COS-7 African green monkey kidney cells.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.