Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-BC Monoclonal Antibody (B1.23.2), APC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. p-7090196
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-3166 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-3166 Supplier Invitrogen™ Supplier No. 17593542
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: This B1.23.2 monoclonal antibody recognizes the human Human Leukocyte Antigens (HLA) B and C. Along with HLA-A, these proteins correspond to MHC Class I in humans. HLA-B and HLA-C are expressed on nearly all cell types and heterodimerize with beta-2 microglobulin at the cell surface. These molecular complexes play a critical role in the presentation of endoplasmic reticulum-derived peptides to immune cells. Applications Reported: This B1.23.2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B1.23.2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

HLA-class I antigens are widely distributed on human nucleated cells. The intensity of HLA-ABC maybe altered in pathological states e.g. malignant cells may lose HLA-ABC, whereas hepatocytes in alcoholic hepatitis, biliary cirrhosis and chronic active hepatitis may show enhanced expression of HLA-ABC. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-BC
Applications Flow Cytometry
Classification Monoclonal
Clone B1.23.2
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene HLA-B
Gene Accession No. P01889
Gene Alias AS; B-4901; B-5001; Bw-47; Bw-50; class I MHC, HLA-B antigen; epididymis secretory protein Li 83; HEL-S-83; histocompatibility antigen; HLA -B; HLA B-40011; HLA B44; HLA class I antigen HLA-B; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibility antigen, B-82 alpha chain; HLA locus; HLA molecule; HLAB; HLA-B; HLA-B alpha chain; HLA-B alpha chain (B*2706); HLA-B alpha chain (B*5002); HLA-B alpha-chain; HLA-B*45ZJ; HLA-B15; HLA-B35; HLA-B35 antigen; HLA-B-3506; HLA-B39; HLA-B-3905; HLA-B49; HLA-B50; HLA-B55; HLA-B-5502; HLA-B-5602; HLA-B59; HLA-B61; HLA-Bw50 antigen; HLA-Cw; HLA-DRB1; hypothetical protein; lecocyte antigen B; Leukocyte Antigen; leukocyte antigen class I-B; Leukoocyte Antigen; lymphocyte antigen; major histocompatability complex B antigen; major histocompatibility complex, class I, B; MHC antigen; MHC class 1 antigen; MHC class I alpha chain; MHC class I anitgen; MHC class I antigen B*47; MHC class I antigen B*50; MHC class I antigen B*82; MHC class I antigen GN00104; MHC class I antigen HLA-B alpha chain; MHC class I antigen HLA-B heavy chain; MHC class I antigen SHCHA; MHC class I antigene; MHC class I human leukocyte antigen; MHC class I molecule; MHC class I protein; MHC class II antigen; MHC class l antigen; MHC classI antigen; MHC HLA-B cell surface glycoprotein; MHC HLA-B transmembrane glycoprotein; MHC HLA-B27d; MHC HLA-B27-HS; MHC HLA-B39N; SPDA1; truncated class I antigen; truncated MHC class I antigen
Gene Symbols HLA-B
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3106
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.