Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-DQ1/DQ3 Monoclonal Antibody (HL-37)
GREENER_CHOICE

Catalog No. 01675915
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-915 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-915 Supplier Invitrogen™ Supplier No. MA119143
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with extracellular polymorphic determinant on human HLA-DQ1 and HLA-DQ3 molecules (recognized epitope was found on isolated beta chain of DQ1), but does not react with HLA-DQ2. DQ is the isotype of human MHC Class II molecules expressed on antigen-presenting cells (APC; dendritic cells, B lymphocytes, monocytes, macrophages). Western Blot: non-reducing conditions.

Reacts with polymorphic determinant on human HLA-DQ1 and HLA-DQ3 molecules (recognized epitope was found on isolated b chain of DQ1), but does not react with HLA-DQ2. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DQ1/DQ3
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HL-37
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene HLA-DQB1
Gene Accession No. P01920
Gene Alias CELIAC1; HLA class II antigen; HLA class II histocompatibility antigen, DQ beta 1 chain; HLA-D; HLADQ; HLA-DQ; HLADQ1; HLADQ3; HLA-DQB; HLA-DQB1; HLA-DRB1; IDDM1; lymphocyte antigen; major histocompatibility complex, class II, DQ beta 1; MHC Class 2; MHC class II; MHC class II antigen DQB1; MHC class II antigen HLA-DQ-beta-1; MHC class II DQ beta chain; MHC class II HLA-DQ beta glycoprotein; MHC class2 antigen; MHC DQ beta; MHC HLA-D; MHC II HLA
Gene Symbols HLA-DQB1
Host Species Mouse
Immunogen Burkitt's lymphoma cell line Raji.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3119
Target Species Human, Pig
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.