Learn More
Diagnostic Biosystems HLA DR HLA-DR (β chain of DP, DQ & DR) - RUO
Supplier:  Diagnostic Biosystems PDM082R
Encompass_Preferred
Reacts with a common epitope of human major histocopatibility (MHC) class II antigens, HLA-DP, -DQ and -DR. Human MHC Class II antigens are transmembrane glycoprotiens composed of an □ chain (36kDa) and a □ chain (27kDa). They are expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, and thymic epithelial cells and are also present on activated T lymphocytes. Human MHC class II genes are located in the HLA-D region that encodes at least six □ and ten □chain genes. Three loci, DR, DQ, and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides and present them to T-helper cells. They, therefore, have a critical role in the intiation of the immune response. It has been shown that some autoimmune diseases are associated with certain class II alleles.
Catalog No. 50-238-5752
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.