Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HLA-DR Monoclonal Antibody (LN3), Alexa Fluor™ 660, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 606995642
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-699-5642 100 Tests
60-699-5641 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-699-5642 Supplier Invitrogen™ Supplier No. 606995642
  • $464.00
    $348.65 / Each of 1

    Save $115.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The LN3 mAb reacts with the human major histocompatibility complex (MHC) class II, HLA-DR. HLA-DR is expressed on the surface of human antigen presenting cells (APC) including B cells, monocytes, macrophages, DCs, and activated T cells. HLA-DR is a heterodimeric transmembrane protein composed of alpha and beta subunits and plays an important role in the presentation of peptides to CD4+ T lymphocytes. Applications Reported: This LN3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This LN3 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor 660 emits at 690 nm and is intended for use on spectral cytometers where it may be multiplexed with APC, Alexa Fluor 647 and Alexa Fluor 700. Excitation: 662 nm; Emission: 690 nm; Laser: Red Laser

HLA-DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of a 36 kDa alpha chain (DRA) and 27 kDa beta chain (DRB). The alpha chain gene contains 5 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, and exon 4 encodes the transmembrane domain and the cytoplasmic tail. DRA does not have polymorphisms in the peptide binding part and acts as the sole alpha chain for DRB1, DRB3, DRB4 and DRB5. Within the DR molecule the beta chain contains all the polymorphisms specifying the peptide binding specificities. Hundreds of DRB1 alleles have been described and typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. HLA-DR is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides, present them to T-helper cells, and have a critical role in the initiation of the immune response. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-DR
Applications Flow Cytometry
Classification Monoclonal
Clone LN3
Concentration 5 μL/Test
Conjugate Alexa Fluor 660
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CD74
Gene Accession No. P01903, P01911, P04233, P13762, P79483, Q30154
Gene Alias AI323765; BLA-DRB3; BOLA-DRA; BoLA-DR-alpha; BoLA-DRB; BOLA-DRB3; BoLA-DRB3 protein; BoLA-DRB3.2; Bota-DRB01; Bota-DRB02; Bota-DRB04; Bota-DRB07; Bota-DRB21; Bota-DRB22; bovine leukocyte antigen; Cd74; CD74 antigen; CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated); CD74 molecule; Cd74 molecule, major histocompatibility complex, class II invariant chain; class II-associated invariant chain peptide; CLIP; Clone P2-beta-3; DASS-397D15.1; DHLAG; dinucleotide microsatellite; DLA class II histocompatibility antigen, DR-1 beta chain; DLA DRBB1 beta chain; DLA-DR beta; DLA-DRB1; DLA-DRBB1; DR beta-5; DR beta-chain antigen binding domain; DR-16; DR2-beta-2; DR4; DR-4; DR7; DR9; DR-9; DRB; DRB1; DRB1 transplantation antigen; DRB3; DRB4; DRBB1; DR-beta chain; DR-beta chain MHC class II; DRw10; Dw2; DW2.2/DR2.2; E-alpha-f; FLJ51114; FLJ75017; FLJ76359; gamma chain of class II antigens; H-2 class II histocompatibility antigen gamma chain; H-2 class II histocompatibility antigen, E-D alpha chain; H-2 class II histocompatibility antigen, E-K alpha chain; H-2 class II histocompatibility antigen, E-U alpha chain; H-2Ea; H2-Ea; H2-Ea-ps; H2-IE-alpha; histocompatibility 2, class II antigen E alpha; histocompatibility 2, class II antigen E alpha, pseudogene; histocompatibility antigen HLA-DR alpha; histocompatibility complex, class II, DR beta 3; histocompatibility: class II antigens, gamma chain of; HLA class II histocompatibility antigen gamma chain; HLA class II histocompatibility antigen, DR alpha chain; HLA class II histocompatibility antigen, DR beta 3 chain; HLA class II histocompatibility antigen, DR beta 4 chain; HLA class II histocompatibility antigen, DR beta 5 chain; HLA class II histocompatibility antigen, DR-1 beta chain; HLA class II histocompatibility antigen, DR-5 beta chain; HLA class II histocompatibility antigen, DRB1-15 beta chain; HLA class II histocompatibility antigen, DRB1-16 beta chain; HLA class II histocompatibility antigen, DRB1-3 chain; HLA class II histocompatibility antigen, DRB1-7 beta chain; HLA class II histocompatibility antigen, DRB1-9 beta chain; HLADG; HLA-DR antigens-associated invariant chain; HLA-DR1B; HLA-DR3B; HLA-DR4B; HLA-DRA; HLA-DRA1; HLA-DRB; HLA-DRB1; HLA-DRB2; HLA-DRB3; HLA-DRB4; HLA-DRB5; HLA-DR-gamma; human leucocyte antigen DRB1; human leucocyte antigen DRB3; human leucocyte antigen DRB4; human leucocyte antigen DRB5; ia antigen-associated invariant chain; Ia3; Ia-3; Ia-associated invariant chain; Ia-GAMMA; I-E alpha MHC class II; I-Ealpha; II; integral membrane glycoprotein; invariant gamma chain; invariant polypeptide of major histocompatibility complex, class II antigen-associated; INVG34; LA-DRB; leukocyte antigen; leukocyte antigen class II; leukocyte antigen DRB3; lymphocyte antigen DRB1; major histocompatibility complex class II DR-beta chain; major histocompatibility complex, class II, DR alpha; major histocompatibility complex, class II, DR alpha precursor; major histocompatibility complex, class II, DR beta 1; major histocompatibility complex, class II, DR beta 3; major histocompatibility complex, class II, DR beta 4; major histocompatibility complex, class II, DR beta 4 precursor; major histocompatibility complex, class II, DR beta 5; major histocompatibility complex, class II, DRB3; MHC cell surface glycoprotein; MHC class I antigen; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen BoLA-DRB3; MHC class II antigen DR beta 3 chain; MHC class II antigen DRA; MHC class II antigen DRB1*15; MHC class II antigen DRB1*16; MHC class II antigen DRB1*3; MHC class II antigen DRB1*9; MHC class II antigen DRB3; MHC class II antigen DRB4; MHC class II antigen DRB5; MHC class II antigen E alpha; MHC class II antigen HLA-DR-beta; MHC class II DLA DRB1 beta chain; MHC class II DLA-DRB; MHC class II DLA-DR-beta-1; MHC class II DR beta 1; MHC class II DR beta chain; MHC class II DR beta-chain
Gene Symbols CD74, HLA-DRA, HLA-DRB1, HLA-DRB3, HLA-DRB4, HLA-DRB5
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3122, 3123, 3125, 3126, 3127, 972
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.