Learn More
Invitrogen™ HLA-DRA Polyclonal Antibody

Description
Immunogen sequence: AIKEEHVIIQ AEFYLNPDQS GEFMFDFDGD EIFHVDMAKK Highest antigen sequence indentity to the following orthologs: Mouse - 41%, Rat - 83%.
Specifications
Specifications
| Antigen | HLA-DRA |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | HLA-DRA |
| Gene Accession No. | P01903 |
| Gene Alias | histocompatibility antigen HLA-DR alpha; HLA class II histocompatibility antigen, DR alpha chain; HLA-DRA; HLA-DRA1; major histocompatibility complex, class II, DR alpha; MHC cell surface glycoprotein; MHC class II antigen DRA; MLRW |
| Gene Symbols | HLA-DRA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.