Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-DRA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7207622 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-1449 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-1449 Supplier Proteintech Group Inc Supplier No. 172211AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Expression of the invariant chain of the HLA-DR antigen (CD74) by B-lymphocytes begins in the pre-B stage but lost before the final differentiation to plasma cells. This protein is expressed in B-lymphocytes and many B-cell lymphomas.

Specifications

Antigen HLA-DRA
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HLA-DRA
Gene Accession No. P01903
Gene Alias HLA DRA, HLA DRA1, HLA-DRA, MHC class II antigen DRA
Gene Symbols HLA-DRA
Host Species Rabbit
Immunogen HLA-DRA Fusion Protein Ag11044
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3122
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.