Learn More
Invitrogen™ HLA-G Monoclonal Antibody (MEM-G/9), Biotin

Description
This antibody reacts with an extracellular epitope on native form of human HLA-G1 on the cell surface as well as with soluble HLA-G5 isoform in its beta2-microglobulin associated form. Reactivity with HLA-G3 was also reported. It will not cross-react with mouse.
Specifications
Specifications
| Antigen | HLA-G |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | MEM-G/9 |
| Concentration | 1 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | HLA-G |
| Gene Accession No. | P17693 |
| Gene Alias | acrosomal protein SP-10; Acrosomal vesicle protein 1; ACRV1; b2 microglobulin; D11S4365; DADB-15K14.8; HLA class I histocompatibility antigen, alpha chain G; HLA class I molecule; HLA G antigen; HLA-6.0; HLAG; HLA-G; HLA-G histocompatibility antigen, class I, G; LOC768350; major histocompatibility complex, class I, G; MHC class I antigen G; MHC-G; MHCIY; MHC-like class I Y; sHLA-G; Soluble HLA class I histocompatibility antigen, alpha chain G; SP-10; SPACA2; sperm protein 10 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.