Learn More
Apexbio Technology LLC Drospirenone 67392-87-4 100mg
SDP
Supplier:  Apexbio Technology LLC B1520100
Encompass_Preferred
Drospirenone (67392-87-4) is a synthetic steroidal progestin targeting the progesterone receptor (PR) and androgen receptor (AR) It is designed to modulate hormonal receptor activity thereby regulating reproductive functions and electrolyte balance Drospirenone exerts its biological activity primarily through progesterone receptor agonism and mineralocorticoid antagonism In experimental studies drospirenone can reduce plasminogen activator inhibitor-1 and tissue plasminogen activator levels via AR interaction Additionally it induces DNA damage in bone marrow cells from female mice Based on these pharmacological properties drospirenone holds research potential in reproductive endocrinology contraception mechanisms progesterone receptor signaling and fluid homeostasis studies
Catalog No. 50-000-87678