Learn More
Apexbio Technology LLC Peramivir Trihydrate 1041434-82-5 50mg
SDP
Supplier:  Apexbio Technology LLC B214750
Encompass_Preferred
Peramivir Trihydrate (CAS 1041434-82-5) is a small-molecule inhibitor targeting influenza viral neuraminidase It is designed to inhibit neuraminidase activity thereby impeding viral particle release and the spread of infection Peramivir Trihydrate exerts its biological activity primarily through competitive inhibition of neuraminidase as a transition-state analog In experimental studies Peramivir Trihydrate demonstrates neuraminidase inhibitory activity with an IC50 of approximately 0 09 nM Based on these pharmacological properties Peramivir Trihydrate holds research potential in investigating influenza virus replication antiviral efficacy and neuraminidase-mediated viral dissemination
Catalog No. 50-000-92294