Learn More
Essendant Hydrogen-Peroxide Cleaner/Disinfectant, 32oz Spray Bottle
Supplier:  Essendant CLO30828/DEL
Encompass_Preferred
Clorox Healthcare Hydrogen Peroxide Cleaner Disinfectant kills microorganisms quickly on a broad range of surfaces. It is a ready-to-use one-step noncorrosive disinfectant cleaner that cleans and disinfects faster than the competition. It has 30 to 60 second hard surface disinfecting times on most bacteria and viruses. On soft surfaces like, curtains, chairs and couches, it kills bacteria in just 30 seconds. It is safe to use on several types of fabrics such as polyester and cotton and it will not harm canvas, nylon or polypropylene. It is EPA-registered to kill pathogens on both hard and soft surfaces. Conscious of patient and staff comfort, this cleaner it is formulated with no harsh chemical odors, fumes, or added fragrances. Intended for use in commercial settings.
Catalog No. NC0349567
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.