Learn More
Epredia™ HM 525 NX Cryostat Blade Holders
Catalog No. 1407039 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Disposable Blade Carrier EC
Disposable Blade Carrier EC70
Standard Knife Holder
For Use With (Equipment):
HM 525 NX and HM 550
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
14-070-39 Disposable Blade Carrier EC HM 525 NX and HM 550
14-070-38 Standard Knife Holder HM 525 NX and HM 550
70-563-0 Disposable Blade Carrier EC70 HM 525 NX and HM 550
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-070-39 Supplier Epredia™ Supplier No. 705470
Add to Cart
Add to Cart
Use Epredia™ HM 525 NX blade holders with HM 525 NX and other Epredia™ cryostats.
Standard Knife Carrier:
  • Allows for insertion of standard knives, with clearance angle adjustment
  • Can be moved horizontally on precision dovetail guide with quick clamp system
  • Includes anti-roll plate
Disposable Blade Carrier EC:
  • Allows for insertion of high- and low-profile blades, with clearance angle adjustment
  • Can be moved on precision dovetail guide with quick clamp plate
  • Includes anti-roll plate
Disposable Blade Carrier EV:
  • Use with vacutome system
  • Allows for insertion of high- and low-profile disposable blades
  • Includes a vacuum anti-roll hood for stretching and disposing of section waste
  • Features a slot that is opened and closed by a vacuum-controlled flap
Magnetic Disposable Blade Carrier MV:
  • Use with vacutome system
  • Allows for insertion of disposable blades
  • Includes a vacuum anti-roll hood for stretching and disposing of section waste
  • Features a slot that is opened and closed by a vacuum-controlled flap
Specifications
Product Type Disposable Blade Carrier EC
For Use With (Equipment) HM 525 NX and HM 550
WARNING: Cancer - www.P65Warnings.ca.gov