Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMBA Linker, Advanced ChemTech

Catalog No. 01339260
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-339-260 5 g
01-339-261 25 g
01-339-262 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-339-260 Supplier Advanced Chem Tech Supplier No. RT10355G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3006-96-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
InChI Key WWYFPDXEIFBNKE-UHFFFAOYSA-N
IUPAC Name 4-(hydroxymethyl)benzoic acid
SMILES OCC1=CC=C(C=C1)C(O)=O

Specifications

Color White/Off-White
Grade Biochemical
Quantity 5 g
Chemical Name or Material HMBA Linker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.