Learn More
HMDS 100% LM Prime, Transene
Catalog No. 502744725
Encompass
Change view
Click to view available options
Quantity:
1 gal.
Catalog No. Quantity
50-274-4725 1 gal.
1 options
Catalog No. 50-274-4725 Supplier Thermo Scientific Production Chemicals and Services Supplier No. T0708134GAL
HMDS (1,1,1,3,3,3-hexamethyldisilazane) 99+%, Electronic Grade for the electronics industry
Hexamethyldisilzane LM (HMDS) is an ultra-high purity, low metals adhesion promoter for photoresists and other photolithography applications.
TRUSTED_SUSTAINABILITY
Specifications
For Use With Deactivating and Coating Chromatographic Supports
Quantity 1 gal.
Standard Concentration 98 to 100%