Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ HMG-CoA Reductase, Mouse, Unlabeled, Clone: IgG-A9,

Catalog No. MABS1233 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1233 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1233 Supplier MilliporeSigma™ Supplier No. MABS1233
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

3-hydroxy-3-methylglutaryl-coenzyme A reductase (EC 1.1.1.34; UniProt P00347; also known as HMG-CoA reductase) is encoded by the HMGCR gene (Gene ID 100756363) in Cricetulus griseus (Chinese hamster). HMG-CoA reductase catalyzes the conversion of HMG-CoA to mevalonic acid, the rate-limiting enzyme of the mevalonate-isoprenoid biosynthesis (MIB) pathway responsible for the production of cholesterol and other isoprenoids, as well as farnesyl pyrophosphate (FPP) required for protein prenylation. HMG-CoA reductase is suppressed by cholesterol derived from the internalization and degradation of low density lipoprotein (LDL) via the LDL receptor as well as oxidized cholesterol species. Competitive inhibitors against HMG-CoA reductase induce LDL receptors expression in the liver, which in turn increases the catabolism of plasma LDL and lowers the plasma concentration of cholesterol. HMG-CoA reductase is a multitransmembrane ER protein with an N-terminal sterol-sensing domain (SSD; a.a. 61-218) and a C-terminal catalytic domain (a.a. 450-888) whose activity is inhibited upon SSD binding by cholesterol. Numerous studies have implicated the involvement of HMG-CoA reductase and mevalonate pathway in carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMG-CoA Reductase
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone IgG-A9
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1 in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene HMGCR
Gene Accession No. P00347
Host Species Mouse
Immunogen Membrane preapration from compactin-adapted CHO cells (Liscum, L., et al. (1983). J. Biol. Chem. 258(13) 8450-8455).
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone IgG-A9 targets an epitope within HMC-CoA reductase catalytic domain (a.a. 450 887).
Target Species Hamster
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.