Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGA1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980803
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0803 Supplier Bioss Supplier No. BS1030R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HMGA1 encodes a non-histone protein involved in many cellular processes, including regulation of inducible gene transcription, integration of retroviruses into chromosomes, and the metastatic progression of cancer cells. The encoded protein preferentially binds to the minor groove of A+T-rich regions in double-stranded DNA. It has little secondary structure in solution but assumes distinct conformations when bound to substrates such as DNA or other proteins. The encoded protein is frequently acetylated and is found in the nucleus.

Specifications

Antigen HMGA1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HMGA1
Gene Accession No. P17096
Gene Alias AL023995; HGNC:5010; high mobility group AT-hook 1; High mobility group AT-hook protein 1; high mobility group HMGA1A; high mobility group HMGA1B; high mobility group protein 1A; High mobility group protein A1; High mobility group protein HMG-I/HMG-Y; high mobility group protein I; High mobility group protein R; high-mobility group (nonhistone chromosomal) protein isoforms I and Y; Hmga1; Hmga1a; Hmga1b; Hmgi; HMG-I(Y); Hmgiy; HMG-R; Hmgy; MGC12816; MGC4242; MGC4854; nonhistone chromosomal high-mobility group protein HMG-I/HMG-Y; non-histone protein; OTTHUMP00000016222; OTTHUMP00000016223; OTTHUMP00000016224; OTTHUMP00000039618
Gene Symbols HMGA1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HMGA1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3159
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.