Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMGA2 Polyclonal Antibody

Catalog No. PIPA521320
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521320 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521320 Supplier Invitrogen™ Supplier No. PA521320
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (95%), Rat (92%), Zebrafish (85%), Xenopus laevis (80%), Chicken (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

HMGA2, previously known as HMGI-C, belongs to the group of high mobility chromosomal proteins that have the AT-hook DNA-binding motif. It is highly and ubiquitously expressed during embryogenesis. Chromosomal rearrangements of the HMGA2 gene and overexpression are frequently associated with benign and malignant tumors, respectively. Human and mouse HMGA2 shares 95% amino acid sequence identity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGA2
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7
Gene Hmga2
Gene Accession No. P52926, P52927
Gene Alias 9430083A20Rik; BABL; high mobility group AT-hook 2; high mobility group AT-hook protein 2; High mobility group protein HMGI-C; high mobility group protein I, isoform C; high-mobility group (nonhistone chromosomal) protein isoform I-C; High-mobility group protein HMGI-C; HMGA2; Hmgic; HMGI-C; LIPO; non-histone chromosomal architectural protein HMGI-C; pg; pygmy; STQTL9
Gene Symbols Hmga2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HMGA2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15364, 8091, 84017
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.