Learn More
Aviva Systems Biology Corp HMGB1 ELISA Kit (Human) (OKCD04074) | 96 Wells
SDP
Supplier:  Aviva Systems Biology Corp OKCD04074
Encompass_Preferred
Aviva Systems Biology HMGB1 ELISA Kit (Human) (OKCD04074) is based on standard sandwich enzyme-linked immuno-sorbent assay technology. An antibody specific for HMGB1 has been pre-coated onto a 96-well plate (12 x 8 Well Strips). Standards or test samples are added to the wells, incubated and removed. A biotinylated detector antibody specific for HMGB1 is added, incubated and followed by washing. Avidin-Peroxidase Conjugate is then added, incubated and unbound conjugate is washed away. An enzymatic reaction is produced through the addition of TMB substrate which is catalyzed by HRP generating a blue color product that changes to yellow after adding acidic stop solution. The density of yellow coloration read by absorbance at 450 nm is quantitatively proportional to the amount of sample HMGB1 captured in the well.`
Catalog No. NC1389742
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.