Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HMGCL Recombinant Protein
SDP

Catalog No. NB6021250UG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6021250UG 50 μg
NB60212250U 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB6021250UG Supplier Novus Biologicals™ Supplier No. NBP26021250UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 28-325 of Human HMGCL The Recombinant Human HMGCL Protein is derived from Baculovirus. The Recombinant Human HMGCL Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4) containing 20% glycerol, 1mM DTT. containingno Preservative
Gene ID (Entrez) 3155
Name Recombinant HMGCL Protein
Purification Method >90%, by SDS-PAGE
Quantity 50 μg
Source Baculovirus
Storage Requirements Store at 4°Cshort term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1ug of protein (determined by LAL method)
Gene Alias 3-hydroxy-3-methylglutaryl-CoA lyase, 3-hydroxymethyl-3-methylglutaryl-CoA lyase, EC 4.1.3.4, HLmitochondrial
Gene Symbol HMGCL
Product Type Recombinant Protein
Recombinant Recombinant
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.