Learn More
Innovative Research Inc Hmn Act tPA Kit
Supplier:  Innovative Research Inc IHUTPAKT
Encompass_Preferred
Human Active tPA ELISA Kit - This kit is for the quantitative determination of active tPA in human plasma and other biological fluids. For research use only. Strip well format. Reagents for up to 96 tests. Functionally active tPA will form a covalent complex with biotinylated human PAI-1 which is bound to the avidin coated on the microtiter plate. Complexed tPA will not bind to the PAI-1 and will not be detected by the assay. After appropriate washing steps, polyclonal anti-human tPA primary antibody binds to the captured tPA. Excess antibody is washed away and bound polyclonal antibody is reacted with the secondary antibody conjugated to horseradish peroxidase. Following an additional washing step, TMB is used for color development at 450nm. A standard calibration curve is prepared along with the samples to be measured using dilutions of human tPA. The amount of color development is directly proportional to the concentration of active tPA in the sample.
Catalog No. 50-274-4248
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.