Learn More
Innovative Research Inc Hmn C1q Kit
Supplier:  Innovative Research Inc IHUC1QKT
Encompass_Preferred
Human Complement Component 1q (C1q) ELISA Kit - The Human Complement Component 1q (C1q) ELISA Kit employs a quantitative enzyme immunoassay technique that measures Complement Component 1q in Human samples. Strip well format for up to 96 tests. It is designed for detection of complement C1q in human plasma, serum, milk, urine, saliva, CSF, and cell culture samples. This assay employs a quantitative sandwich enzyme immunoassay technique that measures human complement C1q in less than 4 hours. A polyclonal antibody specific for human complement C1q has been pre-coated onto a 96-well microplate with removable strips. Complement C1q in standards and samples is sandwiched by the immobilized antibody and a biotinylated polyclonal antibody specific for human complement C1q, which is recognized by a streptavidin-peroxidase (SP) conjugate. All unbound material is washed away and a peroxidase enzyme substrate is added. The color development is stopped and the intensity of the color is measured.
Catalog No. 50-274-4350
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.