Learn More
Innovative Research Inc Hmn Inhibin Beta E Kit
Supplier:  Innovative Research Inc IHUINHBEKT
Encompass_Preferred
Human Inhibin Beta E ELISA Kit - This kit is for the quantitative determination of Human Inhibin Beta E in biofluid samples, such as serum, plasma, tissue homogenates, cell lysates, cell culture supernatants and other biological fluids. This is a sandwich ELISA in strip well format with reagents for up to 96 tests . INHbE present in samples reacts with an anti-Human INHbE capture antibody coated and dried on a microtiter plate. After appropriate washing steps, an anti-INHbE detection antibody conjugated to biotin binds to the captured analyte. Excess antibody is washed away and bound antibody is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is directly proportional to the concentration of active INHbE in the samples. A standard calibration curve is prepared along with the samples to be measured using dilutions of Human INHbE. For RUO.
Catalog No. 50-274-4298
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.