Learn More
Innovative Research Inc Hmn RAP Low Endotoxin 1mg
Supplier:  Innovative Research Inc IHURAPLX1MG
Encompass_Preferred
Human Receptor Associated (RAP) Purified Low Endotoxin 1mg - Human Receptor Associated (RAP) Purified Low Endotoxin is recombinantly produced in E. coli and purified using a combination of conventional and affinity chromatography then endotoxin depleted and sterile filtered. This is a frozen liquid buffered in 0.0067M Phosphate; 0.15M NaCl; pH 7.4 and a purity of >95%. This product is useful for experiments involving signaling events and phosphorylation studies, which may be triggered by endotoxin. More Details Source E. coli Purification Chromatographically purified Purity >95% Storage Conditions -70 C Order fulfillment may require an extended lead time due to the manufacturing process for this product. Please inquire with our Customer Support team for details and current lead time.
Catalog No. 50-274-4260
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.