Learn More
Enzo Life Sciences Hydroxy-2-naphthalenylmethylphosphonic acid (5mg). CAS: 132541-52-7
Supplier: Enzo Life Sciences BMLEI2470005
HNMPA is a membrane impermeable inhibitor of insulin receptor tyrosine kinase (IC50 = 100µM); in rat adipocytes (IC50 = 10µM). Inhibits serine and tyrosine autophosphorylation by the human insulin receptor. Purity: ≥98% (HPLC). Solubility: Soluble in 100% ethanol (25mg/ml), DMSO (25ml/ml) or 5% Sodium bicarbonate (25mg/ml). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.