Learn More
Enzo Life Sciences Cytokeratin 1/5/10/14 (human) monoclonal antibody (34betaE12) (0.5ml)
SDP
Supplier:  Enzo Life Sciences ENZC34903
Encompass_Preferred
Clone: 34βE12. Isotype: IgG1. Immunogen: Keratin solubilized from human stratum corneum. Source: Purified from ascites. Species reactivity: Human. Specificity: 34βE12 is specific for "high molecular weight" cytokeratins 1, 5, 10 and 14, corresponding to molecular weights 68, 58, 56.5 and 50 kDa, respectively, which are characteristically found in "complex" epithelium. The antibody reacts with all squamous and ductal epithelium as well as carcinomas such as squamous cell carcinoma and adenocarcinoma of ductal origin (e.g., breast, pancreas) but does not react with sarcoma, melanoma or lymphoma. This antibody reacts with benign small-acinar lesions of the prostate and has been shown to be useful as an adjunct in the identification of prostate carcinoma. Applications: IHC (PS), WB. Purity: Affinity purified. Formulation: Liquid. In PBS containing 0.1% sodium azide and 1% BSA.
Catalog No. 50-201-3366
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.