Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540574
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540574 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540574 Supplier Invitrogen™ Supplier No. PA540574
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: AGETRGGVGV GGGRKKKTRT VFSRSQVFQL ESTFDLKRYL STAERAGLAA Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 91%; Human: 100%; Mouse: 93%; Rabbit: 79%; Rat: 93%; Zebrafish: 86%.

HMX1 acts as a transcriptional antagonist and is part of the Hmx family of homeodomain proteins which are predominately expressed in discrete regions of developing sensory tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMX1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Hmx1
Gene Accession No. Q9NP08
Gene Alias H6; H6 family homeobox 1; H6 homeo box 1; H6 homeobox 1; H6 homeodomain protein; Hmx1; Homeobox protein H6; homeobox protein HMX1; homeobox protein HMX1-like; homeobox transcription factor SOHo; im:7155045; LOW QUALITY PROTEIN: homeobox protein HMX1; nkx5.3; NKX5-3; zgc:172164
Gene Symbols Hmx1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human HMX1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3166
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.