Learn More
Invitrogen™ HMX2 Polyclonal Antibody

Description
This antibody is predicted to react with chicken, human, Xenopus and zebrafish based on sequence homology.
Specifications
Specifications
| Antigen | HMX2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | HMX2 |
| Gene Accession No. | P43687 |
| Gene Alias | H6 family homeobox 2; H6 homeo box 2; H6 homeobox 2; H6L; HMX2; homeo box (H6 family) 2; homeobox protein H6 family member 2; Homeobox protein HMX2; homeobox protein Nkx-5.2; MGC32455; NK-related homeobox; Nkx-5.2; Nkx5-2 |
| Gene Symbols | HMX2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.