Learn More
TARGETMOL CHEMICALS INC HNMPA 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T21761
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HNMPA is a membrane impermeable inhibitor of insulin receptor tyrosine kinase. HNMPA inhibits tyrosine and serine autophosphorylation by the human insulin receptor. HNMPA exhibits no effect on cyclic AMP-dependent protein kinase or protein kinase C activities. purity: 99%
Catalog No. 50-258-1007
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.