Learn More
Invitrogen™ hnRNP A1 Polyclonal Antibody

Description
Recommended positive controls: 293T, H1299, HeLaS3, mouse brain, PC-12. Predicted reactivity: Mouse (100%), Rat (99%), Xenopus laevis (91%), Pig (100%), Chicken (97%), Rhesus Monkey (98%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | hnRNP A1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.46 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Hnrnpa1 |
| Gene Accession No. | P04256, P09651, P49312 |
| Gene Alias | ALS19; ALS20; Fli-2; Hdp; HDP-1; Helix-destabilizing protein; heterogeneous nuclear ribonucleoprotein A1; Heterogeneous nuclear ribonucleoprotein A1, N-terminally processed; heterogeneous nuclear ribonucleoprotein A1B protein; heterogeneous nuclear ribonucleoprotein B2 protein; heterogeneous nuclear ribonucleoprotein core protein A1; hnRNP A1; hnRNP A1-like 3; hnRNP core protein A1; hnRNP core protein A1-like 3; hnRNP I; Hnrnpa1; hnRNP-A1; HNRNPI; HNRNP-I; Hnrpa1; HNRPA1L3; HNRPA1MGC102835; HNRPI; I79_022927; IBMPFD3; MGC102835; MGC128297 protein; nuclear ribonucleoprotein particle A1 protein; pPTB; PTB; PTB-1; PTB2; PTB3; PTB4; PTB-T; putative heterogeneous nuclear ribonucleoprotein A1-like 3; Putative heterogeneous nuclear ribonucleoprotein A1-like protein 3; ROA1; RP11-78J21.1; single-strand DNA-binding protein UP1; single-strand RNA-binding protein; Single-strand-binding protein; Tis; Topoisomerase-inhibitor suppressed; unwinding protein 1; UP 1; UP1 |
| Gene Symbols | Hnrnpa1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.