Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP CL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116648 Supplier Invitrogen™ Supplier No. PA5116648
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: THP-1, MCF7 Immunogen sequence: NLEKIEKEQS KQEVEVKNAK SEEEQSSSSM KKDETHVKME SEGGAEDSAE EGDPLDDDVN EDQGDDQLEL IKDDEKEAEE GEDDRDSTNG QDDS.

May play a role in nucleosome assembly by neutralizing basic proteins such as A and B core hnRNPs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP CL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HNRNPCL1
Gene Accession No. O60812
Gene Alias heterogeneous nuclear ribonucleoprotein C-like 1; hnRNP C-like-1; hnRNP core protein C-like 1; HNRNPCL1; HNRPCL1
Gene Symbols HNRNPCL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human HNRNPCL1 (NP_0010136531).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343069
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.