Learn More
Invitrogen™ hnRNP F/H Monoclonal Antibody (1G11)

Description
Perform heat mediated antigen retrieval with citrate buffer pH 6 before commencing with IHC staining protocol. Western blot detects a band of approximately 53 kDa (hnRNP F), 56 kDa (hnRNP H). Predicted molecular weight: hnRNP F = 49.8 kDa, hnRNP H = 53.9kDa.
Specifications
Specifications
| Antigen | hnRNP F/H |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1G11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | HNRNPH1 |
| Gene Accession No. | O35737, P31943, P52597, Q9Z2X1 |
| Gene Alias | 4833420I20Rik; AA407306; AI642080; E430005G16Rik; heterogeneous nuclear ribonucleoprotein F; Heterogeneous nuclear ribonucleoprotein F, N-terminally processed; Heterogeneous nuclear ribonucleoprotein H; Heterogeneous nuclear ribonucleoprotein H, N-terminally processed; heterogeneous nuclear ribonucleoprotein H1; heterogeneous nuclear ribonucleoprotein H1 (H); heterogeneous nuclear ribonucleoprotein H1 (H) L homeolog; heterogeneous nuclear ribonucleoprotein H1 L homeolog; heterogeneous nuclear ribonucleoprotein H1 (H) S homeolog; heterogeneous nuclear ribonucleoprotein H1 S homeolog; heterogeneous nuclear ribonucleoprotein H1 L homeolog; heterogeneous nuclear ribonucleoprotein H1 S homeolog; heterogeneous nuclear ribonucleoprotein H2 (H'); hnRNP F; HnRNP F protein; hnRNP H; Hnrnpf; hnrnph; hnrnph1; hnrnph1.L; hnrnph1.S; hnrnph1-a; hnrnph1-b; hnrnph2; Hnrpf; HNRPH; hnrph1; hnrph2; mcs94-1; nucleolin-like protein mcs94-1; OK/SW-cl.23; XELAEV_18017116mg; XELAEV_18021523mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.