Learn More
Description
Recommended positive controls: 293T, A431, Hela, HepG2, Mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (81%), Dog (98%), Pig (99%), Rabbit (99%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | hnRNP K |
| Applications | Immunohistochemistry (Paraffin), Western Blot, RNA Immunoprecipitation, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.18 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | HNRNPK |
| Gene Accession No. | P61978, P61979, P61980 |
| Gene Alias | AUKS; CSBP; dC stretch-binding protein; dC-stretch binding protein; FLJ41122; heterogeneous nuclear ribonucleoprotein K; heterogeneous nuclear ribonucleoprotein K S homeolog; hnRNP K; Hnrnpk; hnRNP-K protein; hnrnpk.S; hnrpk; KBBP; NOVA; RP11-575L7.1; transformation upregulated nuclear protein; Transformation up-regulated nuclear protein; TUNP; XELAEV_18010151mg |
| Gene Symbols | HNRNPK |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
