Learn More
Invitrogen™ HnRNP Q Polyclonal Antibody

Description
The antibody detects endogenous levels of total SYNCRIP protein.
Specifications
Specifications
| Antigen | HnRNP Q |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Syncrip |
| Gene Accession No. | O60506, Q7TMK9, Q7TP47 |
| Gene Alias | 2610109K23Rik; 4632417O19Rik; Ab2-339; Glycine- and tyrosine-rich RNA-binding protein; GRYRBP; GRY-RBP; heterogeneous nuclear ribonucleoprotein Q; hnRNP; hnRNP Q; HNRNPQ; hnRNP-Q; HNRPQ; HNRPQ1; liver regeneration-related protein LRRG077; NS1-associated protein 1; NSAP1; Nsap1l; PP68; RRM RNA binding protein GRY-RBP; synaptotagmin binding cytoplasmic RNA interacting protein; synaptotagmin binding, cytoplasmic RNA interacting protein; synaptotagmin-binding, cytoplasmic RNA-interacting protein; Syncrip |
| Gene Symbols | Syncrip |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.