Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ hnRNP U Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143907
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143907 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143907 Supplier Invitrogen™ Supplier No. PA5143907
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human MOLT-4 whole cell, human Daudi whole cell, human HEL whole cell, human U251 whole cell, rat brain tissue, rat C6 whole cell, mouse brain tissue, mouse NIH/3T3 whole cell. IHC: human breast cancer tissue, human gastric carcinoma tissue, human colorectal adenocarcinoma tissue, human esophageal squamous carcinoma tissue, human esophageal squamous carcinoma tissue, human thyroid cancer tissue, human spleen tissue, mouse brain tissue, rat brain tissue. ICC/IF: SiHa cell. Flow: HEL cell, C6 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The protein encoded by this gene is a receptor for activated C-kinase (RACK) protein. The encoded protein has been shown to bind in vitro to activated protein kinase C beta I. In addition, this protein is a cutaneous T-cell lymphoma-associated antigen. Finally, the protein contains a bromodomain and two zinc fingers, and is thought to be a transcriptional regulator. Multiple transcript variants encoding several different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP U
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene Hnrnpu
Gene Accession No. Q00839, Q8VEK3
Gene Alias AA408410; AI256620; AL024194; AL024437; AW557595; C1orf199; C86794; FLJ30202; FLJ37978; GRIP120; Heterogeneous nuclear ribonucleoprotein U; heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A); heterogenous nuclear ribonucleoprotein U; hnRNP U; HNRNPU; HNRNPU antisense RNA 1; HNRNPU-AS1; Hnrpu; NCRNA00201; nuclear matrix protein sp120; Nuclear p120 ribonucleoprotein; p120; p120 nuclear protein; pp120; rCG_20317; SAFA; SAF-A; scaffold attachment factor A; Scaffold-attachment factor A; SN1; SP120; system N1 Na+ and H+-coupled glutamine transporter; transporter protein; transporter protein; system N1 Na+ and H+-coupled glutamine transporter; U21.1; U211
Gene Symbols Hnrnpu
Host Species Rabbit
Immunogen E.coli-derived human hnRNP U/p120/HNRNPU recombinant protein (Position: E262-L563).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117280, 3192, 51810
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.