Learn More
Aviva Systems Biology Corp HNRPDL ANTIBODY 100UG/100UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP40585_T100
This is a rabbit polyclonal antibody against HNRPDL. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human HNRPDL. Alternate names: HNRNP, JKTBP, JKTBP2, laAUF1, HNRPDLApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53210