Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HO-1/HMOX1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2D10A5, Proteintech
SDP

Catalog No. 501737152 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7152 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7152 Supplier Proteintech Group Inc Supplier No. 667431IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a protein serine/threonine kinase that is activated when bound to the GTP-bound form of Rho. The small GTPase Rho regulates formation of focal adhesions and stress fibers of fibroblasts, as well as adhesion and aggregation of platelets and lymphocytes by shuttling between the inactive GDP-bound form and the active GTP-bound form. Rho is also essential in cytokinesis and plays a role in transcriptional activation by serum response factor. This protein, a downstream effector of Rho, phosphorylates and activates LIM kinase, which in turn, phosphorylates cofilin, inhibiting its actin-depolymerizing activity.

Specifications

Antigen HO-1/HMOX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2D10A5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene HMOX1
Gene Accession No. P06762, P09601, P14901, P32394
Gene Alias bK286B10, heme oxygenase (decycling) 1, Heme oxygenase 1, HMOX1, HO, HO 1, HO1, HSP32
Gene Symbols HMOX1
Host Species Mouse
Immunogen HO-1/HMOX1 Fusion Protein Ag21296
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15368, 24451, 3162, 445512
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.