Learn More
Enzo Life Sciences HO-1 monoclonal antibody (HO-1-1) (1mg)
SDP
Supplier:  Enzo Life Sciences ENZABS6631000
Encompass_Preferred
Clone: HO-1-1. Isotype: IgG1κ. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of human HO-1. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat, Dog, Bovine. Applications: Flow Cytometry, ICC, IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-3190
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.