Learn More
Novus Biologicals™ Hoechst 33342 Inhibitor

Description
Biological Activity: Cell permeable fluorescent DNA stain; binds minor groove of AT-rich regions. Used to quantify DNA in viable cells
Storage: Store at 4°C.
Note: Inhibitors are guranteed for 1 year from date of receipt.
Specifications
Specifications
| Description | CAS: 23491-52-3 |
| Formulation | C27H28N6O.3HCl |
| For Use With (Application) | Inhibits apoptosis |
| Quantity | 50 mg |
| Product Type | Inhibitor |
| Molecular Weight (g/mol) | 561.93g/mol |
| Solubility | Soluble to 100mM in water, 50mM in DMSO |
| Content And Storage | -20°C |
| Target | Hoechst 33342 |
| Color | Yellow |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.