Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Hoechst 33342
GSA_VAGREENER_CHOICESDP

Catalog No. 5117
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-17 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 51-17 Supplier Tocris Bioscience™ Supplier No. 5117/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fluorescent dye for staining DNA

Cell permeable fluorescent DNA stain; binds minor groove of AT-rich regions. Used to quantify DNA in viable cells.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 875756-97-1
Molecular Formula C27H31Cl3N6O

Specifications

Quantity 50 mg
Formula Weight 561.93
Physical Form Solid
Color Yellow
Purity >98%
Molecular Weight (g/mol) 561.94
Chemical Name or Material Hoechst 33342
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.