Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ Buffer Dam for SE 600 Series Electrophoresis Unit
SDP

Catalog No. 03500248
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
SE 600 Series Electrophoresis Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
03-500-248 SE 600 Series Electrophoresis Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-500-248 Supplier Hoefer™ Supplier No. SE6032
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Buffer Dam for SE 600 Series Electrophoresis Unit

  • For use with SE 600 Series Electrophoresis Unit

Specifications

For Use With (Equipment) SE 600 Series Electrophoresis Unit
Type Buffer Dam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.