Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ Cams for SE 400 and SE 600 Series Units and SE 6015 Gel Caster
SDP

Catalog No. 03500244
Encompass
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
SE 400, SE 600 Series Vertical Electrophoresis Unit, SE 6015 Gel Caster
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
03-500-244 SE 400, SE 600 Series Vertical Electrophoresis Unit, SE 6015 Gel Caster
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-500-244 Supplier Hoefer™ Supplier No. SE6005L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cams for SE 400 and SE 600 Series Units and SE 6015 Gel Caster

  • For use with SE 400 and SE 600 Series Units and SE 6015 Gel Caster

Specifications

Color Black
For Use With (Equipment) SE 400, SE 600 Series Vertical Electrophoresis Unit, SE 6015 Gel Caster
Type Cams
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.