Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
SDP

Catalog No. 03500176
Encompass
Encompass_Preferred
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Reference
10
15
18
5
Thickness (Metric):
0.07 cm
0.075 cm
0.1 cm
0.15 cm
Volume (Metric) Well:
120μL/ mm
19.5μL/ mm
2.2μL/ mm
2.9μL/ mm
4.4μL/ mm
4.8μL/ mm
51μL/ mm
68μL/ mm
7.2μL/ mm
9.8μL/ mm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells Thickness (Metric) Volume (Metric) Well
03-500-176 1 Prep, 1 Reference 0.15 cm 120μL/ mm
03-500-169 18 0.1 cm 2.9μL/ mm
03-500-166 15 0.07 cm 2.2μL/ mm
03-500-174 1 Prep, 1 Reference 0.07 cm 51μL/ mm
03-500-170 5 0.07 cm 9.8μL/ mm
03-500-163 10 0.075 cm 9.8μL/ mm
03-500-168 15 0.15 cm 4.4μL/ mm
03-500-167 15 0.1 cm 2.9μL/ mm
03-500-165 10 0.15 cm 7.2μL/ mm
03-500-172 5 0.15 cm 19.5μL/ mm
03-500-164 10 0.1 cm 4.8μL/ mm
03-500-175 1 Prep, 1 Reference 0.1 cm 68μL/ mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 03-500-176 Supplier Hoefer™ Supplier No. SE211AR1.5
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

  • Lightweight
  • Virtually unbreakable

Specifications

For Use With (Equipment) SE 250, SE 260, MiniVE™ Vertical Electrophoresis System
Type Comb
Material PTFE
No. of Wells 1 Prep, 1 Reference
Thickness (English) 0.05 in.
Thickness (Metric) 0.15 cm
Volume (Metric) Well 120μL/ mm
Width (English) Well 2.6 in.
Width (Metric) Well 6.8 cm
Depth (English) 0.51 in.
Depth (Metric) 1.3 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.